siRNA Details
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183392.2 | NP_899248.1 | 6, 7 | 1053 | ||
GenBank mRNA | AK087827.1 | - | 6, 7 | 947 | ||
AK088855.1 | - | 6, 7 | 978 | |||
AK133841.1 | - | 6, 7 | 1051 | |||
AK150182.1 | - | 6, 7 | 978 | |||
AK150841.1 | - | 6, 7 | 939 | |||
AK151896.1 | - | 6, 7 | 939 | |||
AK152136.1 | - | 6, 7 | 939 | |||
AK162037.1 | - | 6, 7 | 985 | |||
BC044303.1 | - | 6, 7 | 981 | |||
BC068114.1 | - | 6, 7 | 938 | 5707888 |
