siRNA Details
Gene Symbol: Fermt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198029.2 | NP_932146.2 | 12 | 2282 | ||
GenBank mRNA | AF073489.1 | - | 9 | 1068 | ||
AK030947.1 | - | 11 | 2472 | |||
AK050804.1 | - | 11 | 1430 | |||
BC029093.1 | - | 3 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
