siRNA Details
Gene Symbol: Fermt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198029.2 | NP_932146.2 | 12, 13 | 2318 | ||
GenBank mRNA | AF073489.1 | - | 9, 10 | 1104 | ||
AK030947.1 | - | 11, 12 | 2508 | |||
AK050804.1 | - | 11, 12 | 1466 | |||
BC029093.1 | - | 3, 4 | 437 |

More Information
Other Species Targeted

Gene Ontology Categories:
