siRNA Details
Gene Symbol: Kcnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008430.2 | NP_032456.2 | 3 | 1538 | ||
GenBank mRNA | AF033017.1 | - | 3 | 1468 | ||
AK005326.1 | - | 1 | 277 | |||
AK137224.1 | - | 3 | 1452 | |||
BC003729.1 | - | 3 | 1235 | 3592756 |
