siRNA Details
Gene Symbol: Klc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025358.2 | NP_001020529.2 | 15 | 2077 | ||
NM_001025359.2 | NP_001020530.2 | 15 | 2104 | |||
NM_001025363.2 | NP_001020534.2 | 14 | 2053 | |||
NM_008450.2 | NP_032476.2 | 14 | 2045 | |||
GenBank mRNA | AA059822.1 | - | 1 | 186 | ||
AF055665.1 | - | 14 | 1939 | |||
CJ249676.1 | - | 1 | 151 | |||
CV883867.1 | - | 2 | 311 | |||
W10467.1 | - | 1 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
