siRNA Details
Gene Symbol: Kpna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008467.4 | NP_032493.1 | 7 | 749 | ||
GenBank mRNA | AF020771.1 | - | 7 | 457 | ||
AK147920.1 | - | 7 | 758 | |||
BC026821.1 | - | 7 | 544 | 3672708 | ||
BC052162.1 | - | 7 | 750 | 6510795 | ||
CT010281.1 | - | 7 | 457 |
