siRNA Details
Gene Symbol: PCYT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312673.1 | NP_001299602.1 | 3 | 338 | ||
NM_005017.3 | NP_005008.2 | 4 | 402 | |||
GenBank mRNA | AK309073.1 | - | 3 | 338 | ||
AK309300.1 | - | 3 | 343 | |||
AK313450.1 | - | 3 | 260 | |||
BC046355.1 | - | 4 | 363 | 5744747 | ||
L28957.1 | - | 3 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
