siRNA Details
Gene Symbol: TRIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003852.3 | NP_003843.3 | 2 | 673 | ||
NM_015905.2 | NP_056989.2 | 2 | 673 | |||
GenBank mRNA | AF009353.1 | - | 2 | 446 | ||
AF119042.1 | - | 2 | 574 | |||
AK075306.1 | - | 2 | 673 | |||
AK302679.1 | - | 3 | 546 | |||
AK312610.1 | - | 2 | 801 | |||
BC028689.2 | - | 2 | 571 | 4827559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
