siRNA Details
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141458.1 | - | 7 | 1537 | ||
AK164828.1 | - | 4 | 1222 | |||
M21041.1 | - | 7 | 1538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
