siRNA Details
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008688.3 | NP_032714.1 | 10 | 1500 | ||
NM_026756.2 | NP_081032.2 | 9 | 1423 | |||
GenBank mRNA | AK087539.1 | - | 10 | 1485 | ||
AK147496.1 | - | 9 | 1408 | |||
AK155258.1 | - | 10 | 1502 | |||
AK164934.1 | - | 2 | 118 | |||
U57635.1 | - | 10 | 1440 | |||
Y07692.1 | - | 10 | 1459 | |||
Y07693.1 | - | 10 | 1805 |

More Information
Other Species Targeted

Gene Ontology Categories:
