siRNA Details
Gene Symbol: Orc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271526.1 | NP_001258455.1 | 4 | 555 | ||
NM_008765.3 | NP_032791.1 | 4 | 561 | |||
GenBank mRNA | AK048408.1 | - | 4 | 380 | ||
AK051747.1 | - | 4 | 390 | |||
AK133604.1 | - | 4 | 380 | |||
AK146281.1 | - | 4 | 380 | |||
AK146859.1 | - | 4 | 374 | |||
AK198581.1 | - | 4 | 234 | |||
BC015257.1 | - | 4 | 371 | 4234871 | ||
U27457.1 | - | 4 | 380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
