siRNA Details
Gene Symbol: EIF4G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB210013.1 | - | 9 | 1696 | ||
AF104913.1 | - | 7 | 1178 | |||
AF281070.1 | - | 9 | 1560 | |||
AK096719.1 | - | 8 | 1207 | |||
AK128378.1 | - | 9 | 1306 | |||
AK294567.1 | - | 8 | 1188 | |||
AL832754.1 | - | 6 | 1031 | |||
AY082886.1 | - | 9 | 1560 | |||
BC082764.1 | - | 3 | 715 | |||
BC140892.1 | - | 11 | 1420 | 9021760 | ||
D12686.1 | - | 3 | 1066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
