siRNA Details
Gene Symbol: Irs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010572.2 | NP_034702.2 | 1 | 830 | ||
GenBank mRNA | BC060235.1 | - | 1 | 830 | ||
BC150782.1 | - | 1 | 904 | 9087693 |

More Information
Other Species Targeted

Gene Ontology Categories:
