siRNA Details
Gene Symbol: Itln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010584.3 | NP_034714.1 | 8 | 1123 | ||
GenBank mRNA | AB016496.1 | - | 8 | 1122 | ||
AK007447.1 | - | 8 | 1123 | |||
AY157363.1 | - | 8 | 1112 | |||
AY157364.1 | - | 8 | 1107 | |||
AY619693.1 | - | 8 | 1106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
