siRNA Details
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145831.1 | NP_001139303.1 | 12 | 1562 | ||
NM_001145832.1 | NP_001139304.1 | 8 | 1046 | |||
NM_010631.2 | NP_034761.3 | 10 | 1389 | |||
GenBank mRNA | AF013118.2 | - | 6 | 1025 | ||
AK132895.1 | - | 8 | 1046 | |||
AK135926.1 | - | 5 | 588 | |||
AK166482.1 | - | 12 | 1541 | |||
BC004069.1 | - | 2 | 137 | |||
BC016118.1 | - | 5 | 593 | |||
BC023374.1 | - | 3 | 181 | |||
BC070429.1 | - | 10 | 1389 | 6810893 | ||
D49546.1 | - | 10 | 1404 |
