siRNA Details
Gene Symbol: ATXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127696.1 | NP_001121168.1 | 3, 4 | 340 | ||
NM_001164778.1 | NP_001158250.1 | 4, 5 | 385 | |||
NM_001164781.1 | NP_001158253.1 | 2, 3 | 175 | |||
NM_004993.5 | NP_004984.2 | 4, 5 | 385 | |||
NM_030660.4 | NP_109376.1 | 3, 4 | 220 | |||
NR_028459.1 | 4, 5 | 385 | ||||
NR_028461.1 | 4, 5 | 385 | ||||
GenBank mRNA | AB050194.1 | - | 3, 4 | 209 | ||
AK225884.1 | - | 4, 5 | 324 | |||
AK314919.1 | - | 4, 5 | 337 | |||
BC033711.1 | - | 4, 5 | 328 | 4393766 | ||
BC095402.1 | - | 4, 5 | 345 | 30347215 | ||
BU190081.1 | - | 3, 4 | 319 | |||
DA827537.1 | - | 4, 5 | 385 | |||
GQ176433.1 | - | 4, 5 | 338 | |||
GQ176434.1 | - | 4, 5 | 337 | |||
GQ176435.1 | - | 4, 5 | 338 | |||
GQ176440.1 | - | 3, 4 | 173 | |||
GQ176452.1 | - | 3, 4 | 338 | |||
GQ176454.1 | - | 4, 5 | 338 | |||
GQ176455.1 | - | 3, 4 | 173 | |||
GQ176462.1 | - | 4, 5 | 338 | |||
GQ176467.1 | - | 3, 4 | 173 | |||
GQ176486.1 | - | 3, 4 | 293 | |||
GQ176488.1 | - | 4, 5 | 338 | |||
GQ176489.1 | - | 3, 4 | 293 | |||
GQ176494.1 | - | 4, 5 | 337 | |||
GQ176499.1 | - | 4, 5 | 338 | |||
GQ176504.1 | - | 4, 5 | 338 | |||
GQ176508.1 | - | 4, 5 | 338 | |||
GQ176510.1 | - | 4, 5 | 338 | |||
GQ176512.1 | - | 4, 5 | 335 | |||
GQ176516.1 | - | 4, 5 | 338 | |||
GQ176523.1 | - | 4, 5 | 338 | |||
GQ176526.1 | - | 2, 3 | 128 | |||
GQ176531.1 | - | 4, 5 | 338 | |||
GQ176532.1 | - | 3, 4 | 338 | |||
GQ176533.1 | - | 4, 5 | 338 | |||
GQ176536.1 | - | 3, 4 | 173 | |||
GQ176538.1 | - | 4, 5 | 338 | |||
GQ176539.1 | - | 3, 4 | 173 | |||
GQ176540.1 | - | 3, 4 | 226 | |||
GQ176548.1 | - | 4, 5 | 338 | |||
GQ176550.1 | - | 4, 5 | 338 | |||
GQ176551.1 | - | 4, 5 | 337 | |||
GQ176552.1 | - | 4, 5 | 337 | |||
GQ176560.1 | - | 4, 5 | 338 | |||
GQ176563.1 | - | 4, 5 | 338 | |||
GQ176565.1 | - | 4, 5 | 338 | |||
GQ176566.1 | - | 3, 4 | 173 | |||
GQ176568.1 | - | 4, 5 | 338 | |||
GQ176570.1 | - | 4, 5 | 338 | |||
GQ176573.1 | - | 3, 4 | 173 | |||
GQ176575.1 | - | 4, 5 | 338 | |||
GQ176576.1 | - | 4, 5 | 338 | |||
GQ176577.1 | - | 4, 5 | 338 | |||
GQ176578.1 | - | 4, 5 | 338 | |||
GQ176580.1 | - | 4, 5 | 338 | |||
GQ176581.1 | - | 338 | ||||
GQ176592.1 | - | 3, 4 | 173 | |||
GQ176593.1 | - | 4, 5 | 337 | |||
GQ176594.1 | - | 4, 5 | 338 | |||
GQ176597.1 | - | 4, 5 | 338 | |||
GQ176599.1 | - | 4, 5 | 338 | |||
GQ176602.1 | - | 4, 5 | 356 | |||
GQ176609.1 | - | 338 | ||||
GQ176612.1 | - | 4, 5 | 338 | |||
GQ176615.1 | - | 338 | ||||
GQ176616.1 | - | 4, 5 | 338 | |||
GQ176618.1 | - | 164 | ||||
GQ176619.1 | - | 4, 5 | 338 | |||
GQ176621.1 | - | 4, 5 | 338 | |||
GQ176622.1 | - | 4, 5 | 338 | |||
GQ176628.1 | - | 338 | ||||
GQ176632.1 | - | 4, 5 | 338 | |||
GQ176640.1 | - | 4, 5 | 338 | |||
GQ176642.1 | - | 4, 5 | 338 | |||
GQ176644.1 | - | 4, 5 | 333 | |||
GQ176645.1 | - | 4, 5 | 338 | |||
GQ176646.1 | - | 4, 5 | 338 | |||
GQ176658.1 | - | 4, 5 | 338 | |||
GQ176660.1 | - | 3, 4 | 252 | |||
GQ176665.1 | - | 3, 4 | 173 | |||
GQ176668.1 | - | 4, 5 | 335 | |||
GQ176671.1 | - | 338 | ||||
GQ176673.1 | - | 4, 5 | 338 | |||
GQ176674.1 | - | 4, 5 | 338 | |||
GQ176675.1 | - | 3, 4 | 173 | |||
GQ176677.1 | - | 4, 5 | 338 | |||
GQ176679.1 | - | 3, 4 | 173 | |||
GQ176680.1 | - | 4, 5 | 338 | |||
GQ176684.1 | - | 4, 5 | 338 | |||
GQ176685.1 | - | 4, 5 | 337 | |||
GQ176686.1 | - | 338 | ||||
GQ176687.1 | - | 4, 5 | 338 | |||
GQ176692.1 | - | 4, 5 | 338 | |||
GQ176693.1 | - | 338 | ||||
GQ176694.1 | - | 3, 4 | 173 | |||
GQ176697.1 | - | 4, 5 | 338 | |||
GQ176698.1 | - | 338 | ||||
GQ176699.1 | - | 338 | ||||
GQ176700.1 | - | 3, 4 | 173 | |||
GQ176701.1 | - | 4, 5 | 338 | |||
GQ176703.1 | - | 4, 5 | 338 | |||
GQ176704.1 | - | 4, 5 | 338 | |||
GQ176708.1 | - | 4, 5 | 338 | |||
GQ176712.1 | - | 3, 4 | 293 | |||
GQ176714.1 | - | 4, 5 | 338 | |||
GQ176715.1 | - | 3, 4 | 173 | |||
GQ176719.1 | - | 338 | ||||
GQ176721.1 | - | 4, 5 | 338 | |||
GQ176725.1 | - | 4, 5 | 338 | |||
GQ176735.1 | - | 4, 5 | 338 | |||
GQ176737.1 | - | 4, 5 | 338 | |||
GQ176741.1 | - | 338 | ||||
GQ176744.1 | - | 3, 4 | 293 | |||
GQ176745.1 | - | 338 | ||||
GQ176746.1 | - | 4, 5 | 338 | |||
GQ176747.1 | - | 338 | ||||
GQ176749.1 | - | 4, 5 | 338 | |||
GQ176750.1 | - | 3, 4 | 173 | |||
GQ176754.1 | - | 3, 4 | 173 | |||
GQ176756.1 | - | 4, 5 | 338 | |||
GQ176759.1 | - | 4, 5 | 338 | |||
GQ176763.1 | - | 4, 5 | 338 | |||
GQ176766.1 | - | 4, 5 | 338 | |||
GQ176776.1 | - | 338 | ||||
GQ176787.1 | - | 4, 5 | 338 | |||
GQ176791.1 | - | 4, 5 | 338 | |||
GQ176800.1 | - | 338 | ||||
GQ176805.1 | - | 4, 5 | 314 | |||
GQ176806.1 | - | 4, 5 | 337 | |||
GQ176808.1 | - | 338 | ||||
GQ176813.1 | - | 3, 4 | 173 | |||
GQ176814.1 | - | 3, 4 | 172 | |||
GQ176820.1 | - | 4, 5 | 338 | |||
GQ176822.1 | - | 3, 4 | 173 | |||
GQ176824.1 | - | 4, 5 | 338 | |||
GQ176825.1 | - | 338 | ||||
GQ176826.1 | - | 4, 5 | 338 | |||
GQ176827.1 | - | 4, 5 | 338 | |||
GQ176829.1 | - | 4, 5 | 338 | |||
GQ176830.1 | - | 309 | ||||
GQ176831.1 | - | 338 | ||||
GQ176835.1 | - | 3, 4 | 172 | |||
GQ176836.1 | - | 3, 4 | 173 | |||
GQ176837.1 | - | 4, 5 | 338 | |||
GQ176838.1 | - | 4, 5 | 338 | |||
GQ176841.1 | - | 4, 5 | 338 | |||
GQ176846.1 | - | 4, 5 | 338 | |||
S75313.1 | - | 4, 5 | 351 | |||
U64820.1 | - | 4, 5 | 373 | |||
U64821.1 | - | 4, 5 | 336 | |||
U64822.1 | - | 3, 4 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
