siRNA Details
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010718.4 | NP_034848.1 | 4 | 530 | ||
NM_173053.1 | NP_774958.1 | 3 | 661 | |||
GenBank mRNA | AB005131.1 | - | 4 | 383 | ||
AB005132.1 | - | 3 | 645 | |||
AB008117.1 | - | 4 | 466 | |||
AK146099.1 | - | 3 | 619 | |||
AK188227.1 | - | 2 | 235 | |||
AK209388.1 | - | 3 | 340 | |||
BC007129.1 | - | 4 | 473 | 3496836 | ||
BC086629.1 | - | 3 | 593 | |||
CV784400.1 | - | 3 | 661 | |||
U88618.1 | - | 4 | 383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
