siRNA Details
Gene Symbol: Il1rl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294171.1 | NP_001281100.1 | 8 | 1181 | ||
NM_010743.3 | NP_034873.2 | 8 | 1348 | |||
GenBank mRNA | AK033417.1 | - | 8 | 1201 | ||
BC051445.1 | - | 8 | 1158 | 30073421 | ||
M24843.1 | - | 8 | 1189 | |||
Y07519.1 | - | 8 | 1192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
