siRNA Details
Gene Symbol: Il1rl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294171.1 | NP_001281100.1 | 8 | 1735 | ||
NM_010743.3 | NP_034873.2 | 8 | 1902 | |||
GenBank mRNA | AK033417.1 | - | 8 | 1755 | ||
M24843.1 | - | 8 | 1743 | |||
Y07519.1 | - | 8 | 1746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
