siRNA Details
Gene Symbol: Mbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025245.1 | NP_001020416.1 | 4 | 521 | ||
NM_010777.3 | NP_034907.1 | 4 | 521 | |||
GenBank mRNA | AK039047.1 | - | 4 | 462 | ||
AK040716.1 | - | 4 | 420 | |||
BC051094.1 | - | 4 | 452 | 3982852 | ||
L07507.1 | - | 4 | 481 | |||
L07508.1 | - | 4 | 498 | |||
X67319.1 | - | 4 | 393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
