siRNA Details
Gene Symbol: Mbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025251.2 | NP_001020422.1 | 7 | 1628 | ||
NM_001025254.2 | NP_001020425.1 | 6 | 1550 | |||
NM_001025255.2 | NP_001020426.1 | 6 | 1550 | |||
NM_001025256.2 | NP_001020427.1 | 6 | 1505 | |||
NM_001025258.2 | NP_001020429.1 | 5 | 1517 | |||
NM_001025259.2 | NP_001020430.1 | 5 | 1427 | |||
NM_010777.3 | NP_034907.1 | 7 | 1973 | |||
GenBank mRNA | AK002947.1 | - | 1 | 95 | ||
AK005129.1 | - | 5 | 1360 | |||
AK079621.1 | - | 6 | 1461 | |||
AK147966.1 | - | 5 | 1375 | |||
AK191097.1 | - | 1 | 48 | |||
AK191806.1 | - | 1 | 48 | |||
AK192498.1 | - | 1 | 48 | |||
AK203705.1 | - | 1 | 48 | |||
BC004704.1 | - | 6 | 1512 | 3499588 | ||
L07507.1 | - | 7 | 1932 | |||
M15060.1 | - | 6 | 1442 | |||
M15062.1 | - | 5 | 1392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
