siRNA Details
Gene Symbol: LIMK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031801.1 | NP_001026971.1 | 3 | 606 | ||
NM_005569.3 | NP_005560.1 | 4 | 459 | |||
NM_016733.2 | NP_057952.1 | 3 | 606 | |||
GenBank mRNA | AB451269.1 | - | 4 | 314 | ||
AB451396.1 | - | 4 | 314 | |||
AK291640.1 | - | 4 | 428 | |||
AK293774.1 | - | 4 | 529 | |||
AK299112.1 | - | 3 | 631 | |||
AK314391.1 | - | 3 | 584 | |||
AL117466.1 | - | 3 | 566 | |||
BC013051.1 | - | 3 | 481 | 3861412 | ||
BU553494.1 | - | 3 | 606 | |||
BX385507.2 | - | 4 | 459 | |||
CR456513.1 | - | 4 | 347 | |||
D45906.1 | - | 4 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
