siRNA Details
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004881.4 | NP_004872.2 | 3, 4 | 1516 | ||
NM_147184.3 | NP_671713.1 | 4, 5 | 1117 | |||
XM_005264650.2 | XP_005264707.1 | 4, 5 | 989 | |||
GenBank mRNA | AF010309.1 | - | 3, 4 | 1154 | ||
AK223382.1 | - | 3, 4 | 1353 | |||
AK297579.1 | - | 3, 4 | 957 | |||
BC000474.2 | - | 4, 5 | 687 | 2961571 | ||
BC018819.2 | - | 4, 5 | 687 | |||
BT007149.1 | - | 3, 4 | 627 |
