siRNA Details
Gene Symbol: Mup3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039544.1 | NP_001034633.1 | 3 | 383 | ||
GenBank mRNA | AI046324.1 | - | 3 | 441 | ||
BC019965.1 | - | 3 | 345 | 4194688 | ||
BC059102.1 | - | 3 | 360 | 4195272 | ||
BC106100.1 | - | 3 | 349 | 5099753 | ||
EU882235.1 | - | 3 | 302 | |||
M27608.1 | - | 3 | 332 | |||
X03525.1 | - | 3 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
