siRNA Details
Gene Symbol: Myh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164171.1 | NP_001157643.1 | 11 | 1131 | ||
NM_010856.4 | NP_034986.1 | 11 | 1067 | |||
GenBank mRNA | AK142232.1 | - | 11 | 1104 | ||
BC061145.1 | - | 11 | 1100 | |||
BC090979.1 | - | 11 | 1100 | |||
BC110700.1 | - | 11 | 1113 | 6786621 | ||
M76599.1 | - | 9 | 929 | |||
M76600.1 | - | 9 | 929 | |||
M76601.1 | - | 11 | 1031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
