siRNA Details
Gene Symbol: Naip7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021545.1 | NP_067520.1 | 4645 |
Gene Symbol: Naip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010871.2 | NP_035001.2 | 14 | 4515 | ||
GenBank mRNA | AF135494.1 | - | 4639 | |||
AF381772.1 | - | 14 | 4515 | |||
AK136436.1 | - | 6 | 1778 | |||
BC116347.2 | - | 13 | 2384 | |||
BC141346.1 | - | 15 | 4581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
