siRNA Details
Gene Symbol: VPS26A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318944.1 | NP_001305873.1 | 9 | 985 | ||
NM_001318945.1 | NP_001305874.1 | 9 | 955 | |||
NM_001318946.1 | NP_001305875.1 | 7 | 803 | |||
NM_004896.4 | NP_004887.2 | 8 | 879 | |||
GenBank mRNA | AF054179.1 | - | 8 | 833 | ||
AF175266.1 | - | 8 | 833 | |||
AK022992.1 | - | 8 | 809 | |||
AK290536.1 | - | 7 | 709 | |||
AK297142.1 | - | 9 | 924 | |||
AK315496.1 | - | 8 | 853 | |||
BC022505.1 | - | 8 | 866 | 4799288 | ||
BX647725.1 | - | 7 | 724 | |||
BX649092.1 | - | 7 | 778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
