siRNA Details
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159717.1 | NP_001153189.1 | 7 | 675 | ||
NM_001159718.1 | NP_001153190.1 | 6 | 648 | |||
NM_001159719.1 | NP_001153191.1 | 7 | 723 | |||
NM_010891.2 | NP_035021.1 | 7 | 679 | |||
GenBank mRNA | AK028072.1 | - | 7 | 661 | ||
AK146616.1 | - | 7 | 676 | |||
AK151591.1 | - | 7 | 663 | |||
AK169852.1 | - | 6 | 697 | |||
AK171331.1 | - | 7 | 658 | |||
BC138636.1 | - | 6 | 616 | 8861656 | ||
BC138637.1 | - | 6 | 616 | 8861657 | ||
D49382.2 | - | 7 | 634 |
