siRNA Details
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037177.2 | NP_001032254.1 | 1, 2 | 302 | ||
NM_001136073.2 | NP_001129545.1 | 1, 2 | 302 | |||
NM_010899.3 | NP_035029.2 | 1, 2 | 302 | |||
GenBank mRNA | AF289078.1 | - | 1, 2 | 128 | ||
AK081853.1 | - | 1, 2 | 269 | |||
AK135414.1 | - | 1, 2 | 303 | |||
BC064006.1 | - | 1, 2 | 221 | 30290121 | ||
CB590507.1 | - | 245 | ||||
EU887582.1 | - | 1, 2 | 128 | |||
EU887588.1 | - | 1, 2 | 128 | |||
EU887594.1 | - | 1, 2 | 128 | |||
EU887600.1 | - | 1, 2 | 128 | |||
U36575.1 | - | 1, 2 | 198 | |||
U36576.1 | - | 1, 2 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
