siRNA Details
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037177.2 | NP_001032254.1 | 2 | 446 | ||
NM_001136073.2 | NP_001129545.1 | 2 | 446 | |||
NM_001291168.1 | NP_001278097.1 | 2 | 331 | |||
NM_001291169.1 | NP_001278098.1 | 2 | 331 | |||
NM_001291172.1 | NP_001278101.1 | 2 | 331 | |||
NM_010899.3 | NP_035029.2 | 2 | 446 | |||
GenBank mRNA | AF289078.1 | - | 2 | 272 | ||
AK044489.1 | - | 2 | 332 | |||
AK081853.1 | - | 2 | 413 | |||
AK135414.1 | - | 2 | 447 | |||
BC064006.1 | - | 2 | 365 | 30290121 | ||
CB590507.1 | - | 389 | ||||
EU887581.1 | - | 2 | 212 | |||
EU887582.1 | - | 2 | 272 | |||
EU887587.1 | - | 2 | 212 | |||
EU887588.1 | - | 2 | 272 | |||
EU887593.1 | - | 2 | 212 | |||
EU887594.1 | - | 2 | 272 | |||
EU887599.1 | - | 2 | 212 | |||
EU887600.1 | - | 2 | 272 | |||
U36575.1 | - | 2 | 342 | |||
U36576.1 | - | 2 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
