siRNA Details
Gene Symbol: Nfatc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136073.2 | NP_001129545.1 | 9 | 2681 | ||
NM_001291168.1 | NP_001278097.1 | 9 | 2566 | |||
NM_001291169.1 | NP_001278098.1 | 9 | 2566 | |||
NM_001291170.1 | NP_001278099.1 | 9 | 2046 | |||
NM_001291171.1 | NP_001278100.1 | 8 | 1530 | |||
NM_001291173.1 | NP_001278102.1 | 8 | 1530 | |||
NM_001291174.1 | NP_001278103.1 | 9 | 2046 | |||
NM_001291175.1 | NP_001278104.1 | 9 | 2161 | |||
NM_001291176.1 | NP_001278105.1 | 8 | 1645 | |||
NM_001291177.1 | NP_001278106.1 | 9 | 2161 | |||
NM_001291178.1 | NP_001278107.1 | 8 | 1645 | |||
NM_010899.3 | NP_035029.2 | 9 | 2681 | |||
GenBank mRNA | AK161174.1 | - | 7 | 1256 | ||
EU887581.1 | - | 9 | 2447 | |||
EU887582.1 | - | 9 | 2507 | |||
EU887583.1 | - | 9 | 2044 | |||
EU887584.1 | - | 9 | 2144 | |||
EU887585.1 | - | 8 | 1528 | |||
EU887586.1 | - | 8 | 1628 | |||
EU887587.1 | - | 9 | 2447 | |||
EU887588.1 | - | 9 | 2507 | |||
EU887589.1 | - | 9 | 2044 | |||
EU887590.1 | - | 9 | 2144 | |||
EU887591.1 | - | 8 | 1528 | |||
EU887592.1 | - | 8 | 1628 | |||
U36575.1 | - | 9 | 2577 | |||
U36576.1 | - | 9 | 2577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
