siRNA Details
Gene Symbol: PDIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK290971.1 | - | 10 | 2340 | ||
BC000425.2 | - | 10 | 2318 | 2819726 | ||
BC001928.1 | - | 10 | 2318 | 2819726 | ||
BC006344.2 | - | 10 | 2507 | 4098367 | ||
BC011754.2 | - | 10 | 2508 | 3532913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
