siRNA Details
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011015.2 | NP_035145.2 | 6 | 876 | ||
GenBank mRNA | AJ003133.1 | - | 6 | 866 | ||
AK042130.1 | - | 6 | 862 | |||
AK141741.1 | - | 6 | 879 | |||
AK145260.1 | - | 6 | 870 | |||
AK161814.1 | - | 6 | 877 | |||
AK164035.1 | - | 6 | 877 | |||
BC015073.1 | - | 6 | 802 | 3995330 | ||
BC038007.1 | - | 5 | 743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
