siRNA Details
Gene Symbol: Ott | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011022.1 | NP_035152.1 | 159 | |||
GenBank mRNA | X96606.1 | - | 159 |
Gene Symbol: Gm10439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037716.2 | NP_001032805.2 | 2 | 128 | ||
GenBank mRNA | BC109132.1 | - | 2 | 128 | 40038983 |
Gene Symbol: Gm15080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198988.1 | NP_001185917.1 | 2 | 159 |
Gene Symbol: Gm15085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122734.1 | NP_001116206.1 | 159 |
Gene Symbol: Gm15091 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122735.1 | NP_001116207.1 | 2 | 159 |
Gene Symbol: Gm15093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099920.1 | NP_001093390.1 | 153 |
Gene Symbol: Gm15097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198987.2 | NP_001185916.2 | 2 | 128 |
Gene Symbol: Gm15107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081648.1 | NP_001075117.1 | 159 | |||
GenBank mRNA | BC109131.1 | - | 128 | 40038981 |
Gene Symbol: Gm15114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082966.1 | NP_001076435.1 | 2 | 153 |
Gene Symbol: Gm15127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114400.2 | NP_001107872.2 | 2 | 153 |
Gene Symbol: Luzp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114383.1 | NP_001107855.1 | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
