siRNA Details
Gene Symbol: DNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005360.2 | NP_001005360.1 | 10 | 1497 | ||
NM_001190716.1 | NP_001177645.1 | 10 | 1497 | |||
NM_004945.3 | NP_004936.2 | 10 | 1497 | |||
GenBank mRNA | AK312260.1 | - | 10 | 1471 | ||
BC039596.1 | - | 10 | 1387 | 5722134 | ||
BC054501.1 | - | 10 | 1457 | 6473791 | ||
L36983.1 | - | 10 | 1451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
