siRNA Details
Gene Symbol: Nodal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013611.4 | NP_038639.2 | 3 | 1822 | ||
GenBank mRNA | AK049290.1 | - | 3 | 1506 | ||
AK082823.1 | - | 3 | 1506 | |||
X70514.1 | - | 3 | 1821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
