siRNA Details
Gene Symbol: Pde1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159582.1 | NP_001153054.1 | 16 | 3006 | ||
NM_016744.4 | NP_058024.2 | 15 | 2801 | |||
GenBank mRNA | AK043647.1 | - | 15 | 2784 | ||
AK134679.1 | - | 1 | 282 | |||
BC090628.1 | - | 16 | 2747 | 6842675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
