siRNA Details
Gene Symbol: NRCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193583.1 | NP_001180512.1 | 9, 10 | 1205 | ||
NM_001193584.1 | NP_001180513.1 | 9, 10 | 1205 | |||
XM_011516262.1 | XP_011514564.1 | 9, 10 | 1205 | |||
XM_017012238.1 | XP_016867727.1 | 9, 10 | 1205 | |||
XM_017012239.1 | XP_016867728.1 | 8, 9 | 1187 | |||
XM_017012240.1 | XP_016867729.1 | 9, 10 | 1205 | |||
XM_017012241.1 | XP_016867730.1 | 8, 9 | 929 | |||
XM_017012242.1 | XP_016867731.1 | 8, 9 | 929 | |||
XM_017012247.1 | XP_016867736.1 | 9, 10 | 1205 | |||
XM_017012249.1 | XP_016867738.1 | 9, 10 | 1205 | |||
XM_017012250.1 | XP_016867739.1 | 8, 9 | 929 | |||
XM_017012255.1 | XP_016867744.1 | 7, 8 | 972 | |||
XM_017012257.1 | XP_016867746.1 | 8, 9 | 960 | |||
GenBank mRNA | AB002341.1 | - | 9, 10 | 1133 | ||
BC114570.1 | - | 9, 10 | 1129 | 40083055 | ||
BC115736.1 | - | 9, 10 | 1129 | 40083062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
