siRNA Details
Gene Symbol: PDE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258312.1 | NP_001245241.1 | 14, 15 | 1790 | ||
NM_005019.4 | NP_005010.2 | 13, 14 | 1805 | |||
XM_017004295.1 | XP_016859784.1 | 14, 15 | 1810 | |||
XM_017004296.1 | XP_016859785.1 | 14, 15 | 1732 | |||
XM_017004297.1 | XP_016859786.1 | 14, 15 | 1673 | |||
XM_017004298.1 | XP_016859787.1 | 14, 15 | 1855 | |||
XM_017004302.1 | XP_016859791.1 | 13, 14 | 1937 | |||
GenBank mRNA | AF110238.1 | - | 2, 3 | 218 | ||
AK294239.1 | - | 14, 15 | 1725 | |||
AK295657.1 | - | 14, 15 | 1595 | |||
BC022480.1 | - | 13, 14 | 1773 | 4814064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
