siRNA Details
Gene Symbol: Meg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018105.1 | - | 5 | 1538 | ||
AK147321.1 | - | 7 | 3005 | |||
BC094421.1 | - | 9 | 1514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
