siRNA Details
Gene Symbol: Myh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039545.2 | NP_001034634.2 | 7 | 760 | ||
GenBank mRNA | BC008538.1 | - | 6 | 699 | 3495412 |
Gene Symbol: Myh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030679.1 | NP_109604.1 | 6 | 719 | ||
GenBank mRNA | AK041122.1 | - | 7 | 732 | ||
BC108329.1 | - | 6 | 719 | 30842458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
