siRNA Details
Gene Symbol: Myh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030679.1 | NP_109604.1 | 19 | 2369 | ||
GenBank mRNA | AK041122.1 | - | 20 | 2382 | ||
BC108329.1 | - | 19 | 2369 | 30842458 | ||
BC150739.1 | - | 18 | 2321 | 9087650 |
Gene Symbol: Myh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039545.2 | NP_001034634.2 | 20 | 2410 | ||
GenBank mRNA | BC008538.1 | - | 19 | 2349 | 3495412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
