siRNA Details
Gene Symbol: ABL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136000.2 | NP_001129472.1 | 3, 4 | 399 | ||
NM_001136001.1 | NP_001129473.1 | 2, 3 | 623 | |||
NM_001168236.1 | NP_001161708.1 | 2, 3 | 623 | |||
NM_001168237.1 | NP_001161709.1 | 3, 4 | 686 | |||
NM_001168238.1 | NP_001161710.1 | 2, 3 | 623 | |||
NM_001168239.1 | NP_001161711.1 | 2, 3 | 336 | |||
NM_005158.4 | NP_005149.4 | 3, 4 | 399 | |||
NM_007314.3 | NP_009298.1 | 3, 4 | 686 | |||
XM_005245088.2 | XP_005245145.1 | 2, 3 | 321 | |||
XM_017001035.1 | XP_016856524.1 | 2, 3 | 2996 | |||
GenBank mRNA | AK225255.1 | - | 2, 3 | 316 | ||
AK309549.1 | - | 3, 4 | 386 | |||
AK311045.1 | - | 2, 3 | 620 | |||
BC065912.1 | - | 3, 4 | 366 | 5173213 | ||
BX538317.1 | - | 3, 4 | 438 | |||
DA596675.1 | - | 3, 4 | 386 | |||
FJ542283.1 | - | 2, 3 | 291 | |||
FJ542284.1 | - | 3, 4 | 399 | |||
FJ542285.1 | - | 2, 3 | 336 | |||
FJ542286.1 | - | 2, 3 | 336 | |||
M35296.1 | - | 3, 4 | 603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
