siRNA Details
Gene Symbol: PRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005039.3 | NP_005030.2 | 2, 3 | 145 | ||
NM_199353.2 | NP_955385.1 | 2, 3 | 145 | |||
NM_199354.2 | NP_955386.1 | 2, 3 | 145 | |||
GenBank mRNA | BC044827.1 | - | 2, 3 | 131 | ||
BC141917.1 | - | 2, 3 | 145 | 40113876 | ||
BG216715.1 | - | 11 | ||||
BX485626.1 | - | 2, 3 | 144 | |||
BX487087.1 | - | 2, 3 | 144 | |||
K03204.1 | - | 2, 3 | 145 | |||
K03205.1 | - | 2, 3 | 133 | |||
K03206.1 | - | 2, 3 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
