siRNA Details
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037997.3 | NP_001033086.2 | 17 | 2305 | ||
NM_001286415.1 | NP_001273344.1 | 16 | 2133 | |||
NM_008000.2 | NP_032026.2 | 12 | 1587 | |||
GenBank mRNA | AK082799.1 | - | 14 | 1798 | ||
BC051249.1 | - | 9 | 1049 | |||
BC058100.1 | - | 16 | 2133 | 6813507 | ||
M32054.1 | - | 12 | 1587 | |||
U76762.1 | - | 16 | 2238 |
