siRNA Details
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010028.3 | NP_034158.1 | 17 | 3859 | ||
GenBank mRNA | BC067210.1 | - | 1 | 642 | ||
BC083059.1 | - | 1 | 650 | |||
U42386.1 | - | 1 | 1462 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108246, XM_006256652, XM_006256653, FQ211849, FQ213286, and others.
Gene Ontology Categories:
