siRNA Details
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010028.3 | NP_034158.1 | 17 | 4021 | ||
GenBank mRNA | AK002917.1 | - | 1 | 89 | ||
BC067210.1 | - | 1 | 804 | |||
BC083059.1 | - | 1 | 812 | |||
U42386.1 | - | 1 | 1624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
