siRNA Details
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135110.2 | NP_001128582.1 | 1, 2 | 296 | ||
NM_005147.5 | NP_005138.3 | 1, 2 | 296 | |||
GenBank mRNA | AB209923.1 | - | 1, 2 | 215 | ||
AF061749.1 | - | 1, 2 | 250 | |||
AK001333.1 | - | 1, 2 | 236 | |||
AK223105.1 | - | 1, 2 | 236 | |||
DA172592.1 | - | 1, 2 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
