siRNA Details
Gene Symbol: H2-T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008208.4 | NP_032234.3 | 2 | 167 | ||
GenBank mRNA | AK033602.1 | - | 2 | 168 | ||
AY989874.1 | - | 2 | 138 | |||
AY989875.1 | - | 2 | 138 | |||
BC040514.1 | - | 178 | ||||
BC125378.1 | - | 2 | 165 | 40129893 | ||
BC125380.1 | - | 2 | 165 | 40129895 | ||
D86082.1 | - | 2 | 144 |
Gene Symbol: H2-T18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153910.1 | - | 2 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
