siRNA Details
Gene Symbol: H2-T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008208.4 | NP_032234.3 | 2 | 315 | ||
GenBank mRNA | AK033602.1 | - | 2 | 316 | ||
AY989874.1 | - | 2 | 286 | |||
AY989875.1 | - | 2 | 286 | |||
BC040514.1 | - | 326 | ||||
BC125378.1 | - | 2 | 313 | 40129893 | ||
BC125380.1 | - | 2 | 313 | 40129895 | ||
D86082.1 | - | 2 | 292 | |||
X03051.1 | - | 1 | 213 |
Gene Symbol: Gm38417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169750.1 | - | 362 |
Gene Symbol: H2-T18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153910.1 | - | 2 | 386 |
Gene Symbol: H2-T3-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY989871.1 | - | 2 | 286 | ||
AY989873.1 | - | 1 | 465 |

More Information
Other Species Targeted

Gene Ontology Categories:
